4-tert-Butyl-1-(bromomethyl)benzene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18880-00-7
:
Formula:C₁₁H₁₅Br
MDL Number:MFCD00000180
:227.14 g/mol
Boiling Pt:93…94 °C (1.5 mmHg)
Melting Pt:8…12 °C
Density:1.236
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:242-643-1
UN:3265
ADR:8,II
:GHS05
Related Information
Frequently Bought Together