
Butylbenzene for synthesis, Sigma-Aldrich®
Catalog # 8.20239.0100
Supplier: Merck
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Solubility:11.8 mg/l (25 °C)
- Melting Point:-88 °C
- Molar Mass:134.22 g/mol
- Boiling Point:183 °C
- Vapor Pressure:1.33 hPa (23 °C)
- Flash Point:58 °C
- Refractive Index:1.4895 (20 °C, 589 nm)
- Explosion Limit:0.8 - 5.8 %(V)
- Density:0.86 g/cm3 (20 °C)
- Ignition Point:410 °C
Specification Test Results
About this item
:
:134.22 g/mol
Merck Index:13,01548
Boiling Pt:183 °C (1013 hPa)
Melting Pt:–88 °C
Density:0.86 g/cm³ (20 °C)
Flash Pt:61 °C
Storage Temperature:Ambient
EINECS:203-209-7
UN:2709
ADR:3,III
:M-Clarity™ quality level = MQ200
:GHS02
CAS Number:104-51-8
Formula:C₁₀H₁₄
MDL Number:MFCD00009463