

α,3,4-Trichlorotoluene
Catalog # EHERC17787800
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:195.47 g/mol
Boiling Pt:241 °C (1013 hPa)
Melting Pt:37.5 °C
Density:1.41 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-033-0
UN:3265
ADR:8,II
:GHS05
:GHS07
CAS Number:102-47-6
Formula:C₇H₅Cl₃
MDL Number:MFCD00000906