Benzyl chloride
Catalog # EHERC11519000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:126.59 g/mol
Merck Index:13,01131
Boiling Pt:179.3 °C (1013 hPa)
Melting Pt:–41.2 °C
Density:1.103 g/cm³ (20 °C)
Flash Pt:52 °C
EINECS:202-853-6
UN:1738
ADR:6.1,II
:GHS06
:GHS05
:GHS08
Storage Temperature:Ambient
CAS Number:100-44-7
Formula:(C₆H₅)CH₂Cl
MDL Number:MFCD00000889