Order Entry
Spain
ContactUsLinkComponent
Pesticide reference standard, 1,4-Dichlorobenzene 10 µg/ml in isooctane, Dr Ehrenstorfer
Pesticide reference standard, 1,4-Dichlorobenzene 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL12372000IO
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, 1,4-Dichlorobenzene 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL12372000IO
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    106-46-7
  • Description:
    1,4-Dichlorobenzene 10 µg/ml in isooctane
  • Formula:
    C₆H₄Cl₂
  • Description:
    1,4-Dichlorobenzene

Specifications

About this item