(R)-(+)-3-Bromo-α-methylbenzyl alcohol 95%, 98% ee
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:134615-24-0
:
Formula:C₈H₉BrO
MDL Number:MFCD06658929
:201.06 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- GC:≥94 %
- Enantiomeric excess:≥97.5 % (GC)
Specification Test Results
Frequently Bought Together