Order Entry
Spain
ContactUsLinkComponent
Erythrosin B disodium salt
Erythrosin B disodium salt
Catalog # 720-1142
Supplier:  RAL
CAS Number:  
Erythrosin B disodium salt
Catalog # 720-1142
Supplier:  RAL
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :879.86 g/mol
EINECS:240-474-8
 :GHS07
 :1
CAS Number:16423-68-0
 :568-63-8
Formula:C₂₀H₆I₄Na₂O₅
MDL Number:MFCD00144257