Order Entry
Spain
ContactUsLinkComponent
Erythrosin B disodium salt, Sigma-Aldrich®
Erythrosin B disodium salt, Sigma-Aldrich®
Catalog # E8886-25G
CAS Number:  
undefined
Erythrosin B disodium salt, Sigma-Aldrich®
Catalog # E8886-25G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :879.86 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:240-474-8
 :GHS07
CAS Number:16423-68-0
 :568-63-8
Formula:C₂₀H₆I₄Na₂O₅
MDL Number:MFCD00144257