Order Entry
Spain
ContactUsLinkComponent
Erythrosin B disodium salt, pure, high purity biological stain
Erythrosin B disodium salt, pure, high purity biological stain
Catalog # ACRO189460250
CAS Number:  
undefined
Erythrosin B disodium salt, pure, high purity biological stain
Catalog # ACRO189460250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Red to bordeaux Powder
  • Infrared spectrum:
    Conforms
  • Dye content:
    ≥87 %
  • Arsenic (As):
    ≤3 ppm
  • Cadmium (Cd):
    ≤1 ppm
  • Lead (Pb):
    ≤10 ppm
  • Mercury (Hg):
    ≤1 ppm
  • Insoluble matter:
    ≤0.2 % (in water)
  • Subsidiary dyes:
    ≤4.0 %
  • Ether extractable matter:
    ≤0.2 %
  • Fluorescein:
    ≤20 ppm
  • Inorganic iodide:
    ≤0.1 % (as Sodium iodide)

Specification Test Results

About this item

CAS Number:16423-68-0
 :568-63-8
Formula:C₂₀H₆I₄Na₂O₅
MDL Number:MFCD00144257
 :879.86 g/mol
Melting Pt:>300 °C
EINECS:240-474-8
 :GHS07
Storage Temperature:Ambient