Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Erythrosin B disodium salt
Erythrosin B disodium salt
Erythrosin B disodium salt
Erythrosin B disodium salt
EHERC13205000
:
EHRENSTORFER
:
Erythrosin B disodium salt
EHERC13205000
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
0,25 g
Seq:
3029
:
:
879.86 g/mol
:
240-474-8
:
GHS07
:
16423-68-0
:
568-63-8
:
C₂₀H₆I₄Na₂O₅
:
MFCD00144257