Order Entry
Spain
ContactUsLinkComponent
Fast green FCF, Sigma-Aldrich®
Fast green FCF, Sigma-Aldrich®
Catalog # F7258-25G
CAS Number:  
undefined
Fast green FCF, Sigma-Aldrich®
Catalog # F7258-25G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :808.86 g/mol
Merck Index:13,03970
Melting Pt:290 °C
 :M-Clarity™ quality level = MQ200
EINECS:219-091-5
 :GHS08
Storage Temperature:Ambient
CAS Number:2353-45-9
Formula:C₃₇H₃₄N₂Na₂O₁₀S₃
MDL Number:MFCD00013053