Order Entry
Spain
ContactUsLinkComponent
Orthoboric acid 4% ± 0.05% in aqueous solution, Fluka™
Health
Orthoboric acid 4% ± 0.05% in aqueous solution, Fluka™
Catalog # 31144-2.5L
CAS Number:  
undefined
Health
Orthoboric acid 4% ± 0.05% in aqueous solution, Fluka™
Catalog # 31144-2.5L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay:
    3.95 - 4.05 %

Specification Test Results

About this item

 :
 :61.83 g/mol
EINECS:233-139-2
 :GHS08
Storage Temperature:Ambient
CAS Number:10043-35-3
Formula:B(OH)₃
MDL Number:MFCD00011337