
2-Fluoro-5-nitroaniline 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:369-36-8
:
Formula:C₆H₅FN₂O₂
MDL Number:MFCD00007652
Melting Pt:99…103 °C
Flash Pt:91 °C
Storage Temperature:Ambient
EINECS:206-720-3
UN:2811
ADR:6.1,III
:156.12 g/mol
:GHS07
Specification Test Results
- Appearance:Ochre to light brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:99 to 103 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together