Order Entry
Spain
ContactUsLinkComponent
 
 
 
 
2-Chlorophenol for synthesis, Sigma-Aldrich®
 :  Merck
 
 
 
 
2-Chlorophenol for synthesis, Sigma-Aldrich®
 :  Merck
 

 

  • Solubility:
    28.5 g/l (20 °C)
  • Melting Point:
    7 °C
  • Molar Mass:
    128.55 g/mol
  • Boiling Point:
    174 °C (1013 hPa)
  • Vapor Pressure:
    13.3 hPa (50 °C)
  • Flash Point:
    63.9 °C
  • Refractive Index:
    1.5579 (20 °C, 589 nm)
  • Density:
    1.26 g/cm3 (20 °C)
  • Ignition Point:
    550 °C
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic Materials

 

Frequently Bought Together