

2-Chlorophenol
Catalog # EHERC11470000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,5 g
- Seq:3035
Specifications
About this item
:
:128.56 g/mol
Merck Index:12,02173
Boiling Pt:175 °C (1013 hPa)
Melting Pt:7-8 °C
Melting Pt:7…8 °C
Density:1.25 g/cm³ (20 °C)
Flash Pt:72 °C
Storage Temperature:Ambient
EINECS:202-433-2
UN:2021
ADR:6.1,III
:GHS07
:GHS09
CAS Number:95-57-8
Formula:C₆H₅ClO
MDL Number:MFCD00002159