2,4-Dichlorophenol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:120-83-2
:
Formula:C₆H₄Cl₂O
MDL Number:MFCD00002169
:163 g/mol
Merck Index:13,03098
Boiling Pt:210 °C (1013 hPa)
Melting Pt:40…41 °C
Density:1.4 g/cm³ (20 °C)
EINECS:204-429-6
UN:2020
ADR:6.1,III
:M-Clarity™ quality level = MQ200
Flash Pt:114 °C
:GHS06
:GHS05
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:4.5 g/l (20 °C)
- Melting Point:40 - 43 °C
- Molar Mass:163 g/mol
- Boiling Point:209 - 211 °C (1013 hPa)
- Vapor Pressure:1.3 hPa (50 °C)
- Flash Point:114 °C
Specification Test Results
Frequently Bought Together