


2,6-Dichloroaniline
Catalog # PROOMM0006.21
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:162.02 g/mol
Boiling Pt:220 °C (1013 hPa)
Melting Pt:35…38 °C
Density:1.526 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-160-5
UN:3442
ADR:6.1,II
:GHS06
:GHS08
:GHS09
CAS Number:608-31-1
Formula:C₆H₅Cl₂N
MDL Number:MFCD00007675