


2,4-Dichloroaniline
Catalog # EHERC12322400
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,5 g
- Seq:3035
Specifications
About this item
:
:162.02 g/mol
Boiling Pt:245 °C (1013 hPa)
Melting Pt:59…62 °C
Density:1.567 g/cm³ (20 °C)
Flash Pt:149 °C
Storage Temperature:Ambient
EINECS:209-057-8
UN:3442
ADR:6.1,II
:GHS06
:GHS08
:GHS09
CAS Number:554-00-7
Formula:C₆H₅Cl₂N
MDL Number:MFCD00007661