Order Entry
Spain
ContactUsLinkComponent
2,3-Dichloroaniline
2,3-Dichloroaniline
Catalog # EHERC12322300
Supplier:  EHRENSTORFER
CAS Number:  
2,3-Dichloroaniline
Catalog # EHERC12322300
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,5 g
  • Seq:
    3035

Specifications

About this item

 :
 :162.02 g/mol
Boiling Pt:252 °C (1013 hPa)
Melting Pt:23…24 °C
Density:1.383 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:210-157-9
UN:3442
ADR:6.1,II
CAS Number:608-27-5
Formula:C₆H₅Cl₂N
MDL Number:MFCD00007657