Order Entry
Spain
ContactUsLinkComponent
3-Chlorobenzoic acid
Irritant
3-Chlorobenzoic acid
Catalog # PROONIST-2144
CAS Number:  
Irritant
3-Chlorobenzoic acid
Catalog # PROONIST-2144
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2 g
  • Seq:
    3107

Specifications

About this item

 :
 :156.57 g/mol
Merck Index:13,02142
Boiling Pt:281 °C (1013 hPa)
Melting Pt:122 °C
Density:1.496 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-618-4
 :GHS07
CAS Number:535-80-8
Formula:C₇H₅ClO₂
MDL Number:MFCD00002491