
3-Chlorobenzoic acid 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:535-80-8
:
Formula:C₇H₅ClO₂
MDL Number:MFCD00002491
:156.57 g/mol
Merck Index:13,02142
Boiling Pt:281 °C (1013 hPa)
Melting Pt:154…157 °C
Density:1,496 g/cm³ (20 °C)
Flash Pt:150 °C
EINECS:208-618-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:154 to 157 °C
- HPLC:≥99.0 %
- Titration with NaOH:99.0 to 100.5 %
- BSC Certification:Passes
Specification Test Results
Frequently Bought Together