Order Entry
Spain
ContactUsLinkComponent
 
2,4-Dichlorobenzonitrile for synthesis, Sigma-Aldrich®
Catalog # 8.40037.0100
Supplier:  Merck
CAS Number:  
2,4-Dichlorobenzonitrile for synthesis, Sigma-Aldrich®
Catalog # 8.40037.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    59 - 62 °C
  • Molar Mass:
    172.01 g/mol

Specification Test Results

About this item

 :
 :172.01 g/mol
Boiling Pt:261 °C (1013 hPa)
Melting Pt:52…53 °C
Storage Temperature:Ambient
ADR:6.1,III
 :M-Clarity™ quality level = MQ200
EINECS:229-493-2
UN:3439
 :GHS07
CAS Number:6574-98-7
Formula:C₇H₃Cl₂N
MDL Number:MFCD00016373