Order Entry
Spain
ContactUsLinkComponent
Pesticide reference standard, 1,4-Dichlorobenzene 1000 µg/ml in methanol, Dr Ehrenstorfer
Pesticide reference standard, 1,4-Dichlorobenzene 1000 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERYA12372000ME
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, 1,4-Dichlorobenzene 1000 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERYA12372000ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    106-46-7
  • Description:
    1,4-Dichlorobenzene 1,000 µg/ml in methanol
  • Formula:
    C₆H₄Cl₂
  • Description:
    1,4-Dichlorobenzene

Specifications

About this item