1,4-Dichlorobenzene 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:106-46-7
:
Formula:C₆H₄Cl₂
MDL Number:MFCD00000604
:147 g/mol
Merck Index:13,03082
Boiling Pt:174 °C (760 mmHg)
Melting Pt:52…56 °C
Density:1.24 g/cm³
Flash Pt:67 °C
Storage Temperature:Ambient
EINECS:203-400-5
UN:3077
ADR:9,III
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:White Crystals
- Infrared spectrum:Conforms
- Melting point:52 to 55 °C
- GC:≥99.0 %
- UV Lambda max:528 nm (in Methanol)
- Fluorescence max:547 nm (in Methanol)
- Optimum lasing:563 nm (in Methanol)
- Suitability:(for use as a laser dye) Passes test
- Molar absorption coefficient:104000 l/(mole.cm) (Lambda max)
Specification Test Results
Frequently Bought Together