1,2-Dichlorobenzene 98% for spectrophotometry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:95-50-1
:
Formula:C₆H₄Cl₂
MDL Number:MFCD00000535
:147 g/mol
Merck Index:14,3056
Boiling Pt:180 °C
Melting Pt:–18 °C
Density:1.306
Flash Pt:68 °C (154 °F)
Storage Temperature:Ambient
EINECS:202-425-9
UN:1591
ADR:6.1,III
:GHS07
:GHS09
Related Information
Frequently Bought Together