
Pyromellitic acid 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:89-05-4
:
Formula:C₁₀H₆O₈
MDL Number:MFCD00002471
:254.15 g/mol
Merck Index:12,08188
Boiling Pt:516 °C (1013 hPa)
Melting Pt:281…284.5 °C
Density:1,79 g/cm³ (20 °C)
EINECS:201-879-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Titration with NaOH:≥95 %
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together