4-Aminobenzoic acid
Catalog # USBIA1375
Supplier: US Biological
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:137.14 g/mol
Merck Index:13,00422
Boiling Pt:340 °C (1013 hPa)
Melting Pt:188 °C
Density:1.45 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:205-753-0
CAS Number:150-13-0
Formula:C₇H₇NO₂
MDL Number:MFCD00007894