Order Entry
Spain
ContactUsLinkComponent
4-Aminobenzoic acid 99%
4-Aminobenzoic acid 99%
Certificates
undefined
4-Aminobenzoic acid 99%
Certificates

Specification Test Results

  • Appearance:
    White to light yellow to light grey to light brown Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    186 to 189 °C
  • Titration with NaOH:
    98.5 to 101.5 %
  • Elemental Analysis:
    13.32 to 14.00 % N

Specification Test Results

Related Information

Frequently Bought Together