

Ácido benzoico 99.5-100.5% (by acidimetry on anhydrous substance), en polvo, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, E210, SAFC®
1.00130.9025
:
- Tipo de env.:Envase de cartón ondulado
- Env:25 kg
- Seq:4283
- Assay (Acidimetric):99.5 - 100.5%
- Assay (Acidimetric, Calculated on Anhydrous Substance):99.5 - 100.5%
- Identity:Conforms
- Appearance of Solution (5%; Ethanol 96%):Passes Test
- pH-Value (Saturated Solution):2.5 - 3.5
- Solidification Point (Lower Value):≥121 °C
- Solidification Point (Upper Value):≤123 °C
- Sulfate (SO₄):≤0.02%
- As (Arsenic):≤1 ppm
- Cd (Cadmium):≤10 ppm
- Cr (Chromium):≤2.5 ppm
- Cu (Copper):≤10 ppm
- Hg (Mercury):≤1 ppm
- Mn (Manganese):≤25 ppm
- Ni (Nickel):≤2 ppm
- Pb (Lead):≤2 ppm
- Zn (Zinc):≤10 ppm
- Halogenated Compounds (as Cl):≤0.01%
- Toluene (HS-GC):≤890 ppm
- Other Residual Solvents (ICH Q3C):Excluded by the Production Process
- Oxidizable Matter (Ph. Eur.):Passes Test
- Oxidizable Matter (USP):Passes Test
- Readily Carbonisable Substance:Passes Test
- Water:≤0.5%
- Sulfated Ash:≤0.05%
:
:122.12 g/mol
:13,01092
:249 °C (1013 hPa)
:121…123 °C
:1,316 g/cm³ (20 °C)
:200-618-2
:01-2119455536-33
:GHS05
:GHS08
:Ambiente
:65-85-0
:C₇H₆O₂
:MFCD00002398