4-Hydroxybenzonitrile 99%
:767-00-0
:
:C₇H₅NO
:MFCD00002312
:148 °C (1 -rr)
:110…113 °C
:~ 1 g/cm³ (20 °C)
:Ambient
:212-175-2
:6.1,III
:119.12 g/mol
:GHS07
- Appearance:Off-white to beige to light grey Crystalline powder
- Infrared spectrum:Conforms
- Melting point:110 to 113 °C
- Water:≤0.4 %
- Sulfated ash:≤0.2 %
- GC:≥98.5 %
Frequently Bought Together