
Benzaldehyde 98+%
:100-52-7
:
:C₆H₅CHO
:MFCD00003299
:106.12 g/mol
:13,01057
:179 °C (1013 hPa)
:–26 °C
:1.043 g/cm³
:64 °C
:Ambient
:202-860-4
:1990
:9,III
:GHS07
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5440 to 1.5460 (20°C, 589 nm)
- GC:≥98.0 %
- Specific gravity:(25°C) 1.041 to 1.046
- Acid value:≤5 mg KOH/g
Frequently Bought Together