1-Bromo-2,4-dichlorobenzene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1193-72-2
:
Formula:C₆H₃BrCl₂
MDL Number:MFCD00018333
:225.9 g/mol
Boiling Pt:235 °C
Melting Pt:26…27 °C
Storage Temperature:Ambient
EINECS:214-778-6
:GHS07
Related Information
Frequently Bought Together