QuEChERS extraction products
The QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) technique is increasingly becoming the technique of choice for the extraction of pesticide residues in fruit, vegetables and other foods. The QuEChERS method offers the advantage of high recoveries, accurate results and high sample throughput, combined with the associated cost savings with low solvent and labour requirements. Yet the procedure is robust and rugged.
- Simple manual procedure
- Determine wide range of pesticide types
- Less solvent usage than conventional techniques
- High recoveries, accurate results and high sample throughput
The QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) technique is increasingly becoming the technique of choice for the extraction of pesticide residues in fruit, vegetables and other foods. The QuEChERS method offers the advantage of high recoveries, accurate results and high sample throughput, combined with the associated cost savings with low solvent and labour requirements. Yet the procedure is robust and rugged.
- Simple manual procedure
- Determine wide range of pesticide types
- Less solvent usage than conventional techniques
- High recoveries, accurate results and high sample throughput
Product Family Options
HyperSep™ dispersive SPE clean-up products
- DescripciónCapacidad
- Column with 250 mg graphatised carbon on top, 500 mg PSA on bottom, separated by a PTFE frit6 ml1 * 30 UN315,00 €
- Tube with 900 mg anhydrous magnesium sulphate, 300 mg PSA and 150 mg endcapped C1815 ml1 * 50 UN