
4-Methyl-1-pentene 97%
Catalog # ACRO127420250
Supplier: Thermo Fisher Scientific
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3810 to 1.3830 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
About this item
:
:84.16 g/mol
Boiling Pt:54 °C (1013 hPa)
Melting Pt:–155 °C
Density:0.66 g/cm³
Flash Pt:–31 °C
Storage Temperature:Refrigerator
EINECS:211-720-1
UN:3295
ADR:3,II
:GHS02
CAS Number:691-37-2
Formula:C₆H₁₂
MDL Number:MFCD00008949