Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fittings and Connectors
Intramedic™ Luer-Stub adapters (sterile)
Intramedic™ Luer-Stub adapters (sterile)
Intramedic™ Luer-Stub adapters (sterile)
Catalog # DIFC427563
Supplier:
DIFCO BD
Expand image
Intramedic™ Luer-Stub adapters (sterile)
Catalog # DIFC427563
Supplier:
DIFCO BD
Specifications
For:
PE190, 160 tubing
Tubing int.Ø:
1,19 mm
Description:
Intramedic™ 18-gauge Luer-Stub adapter
Pk:
100
Specifications
About this item
Product Details
Product Details