Order Entry
Spain
ContactUsLinkComponent
 
 
 
 
 
2-Methylhexane 99%
 
undefined
 
 
 
 
2-Methylhexane 99%

 

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3835 to 1.3855 (20°C, 589 nm)
  • GC:
    ≥98.5 %

 

 

Frequently Bought Together