



2,2,4-Trimethylpentane ≥99.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
:540-84-1
:
:(CH₃)₂CHCH₂C(CH₃)₃
:MFCD00008943
:114.23 g/mol
:13,05212
:99.2 °C (1013 hPa)
:–107 °C
:0.692 g/cm³ (20 °C)
:–12 °C
:208-759-1
:1262
:3,II
:GHS02
:GHS08
:GHS07
:GHS09
:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Assay (on anhydrous substance):Min. 99.5 %
- IR Spectrum:Passes test
- Acidity:Max. 0.0003 meq/g
- Boiling point:99 - 100 °C
- Colouration:Max. 10 APHA
- Density (20/4):0.690 - 0.695
- Density (20/20):0.691 - 0.696
- Refractive index (20°C):1.391 - 1.393
- Substances coloured by H2SO4:Max. 35 APHA
- Evaporation residue:Max. 10 ppm
- Sulphur compounds (as S):Max. 10 ppm
- Water:Max. 100 ppm
- Al (Aluminium):Max. 0.5 ppm
- B (Boron):Max. 0.02 ppm
- Ba (Barium):Max. 0.1 ppm
- Ca (Calcium):Max. 0.5 ppm
- Cd (Cadmium):Max. 0.05 ppm
- Co (Cobalt):Max. 0.02 ppm
- Cr (Chromium):Max. 0.02 ppm
- Cu (Copper):Max. 0.02 ppm
- Fe (Iron):Max. 0.1 ppm
- K (Potassium):Max. 0.1 ppm
- Mg (Magnesium):Max. 0.1 ppm
- Mn (Manganese):Max. 0.02 ppm
- Na (Sodium):Max. 0.5 ppm
- Ni (Nickel):Max. 0.02 ppm
- Pb (Lead):Max. 0.1 ppm
- Sn (Tin):Max. 0.2 ppm
- Sr (Strontium):Max. 0.05 ppm
- Zn (Zinc):Max. 0.1 ppm
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test
Frequently Bought Together