3,3-Dimethylbutanone for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:75-97-8
:
Formula:C₆H₁₂O
MDL Number:MFCD00008846
:100.16 g/mol
Merck Index:12,07594
Boiling Pt:106 °C (1013 hPa)
Melting Pt:–53 °C
Density:0.803 g/cm³ (20 °C)
EINECS:200-920-4
UN:1993
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:5 °C
:GHS02
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:19 g/l (25 °C)
- Melting Point:-53 °C
- Molar Mass:100.16 g/mol
- Boiling Point:105 - 106 °C (1013 hPa)
- Vapor Pressure:33 hPa (20 °C)
- Flash Point:5 °C
- Density:0.808 g/cm3 (20 °C)
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Specification Test Results
Frequently Bought Together