

Isobutyraldehyde for synthesis, Sigma-Aldrich®
: Merck
:78-84-2
:
:C₄H₈O
:MFCD00006980
:72.11 g/mol
:13,05171
:64 °C (1013 hPa)
:–65 °C
:0.794 g/cm³ (20 °C)
:201-149-6
:2045
:3,II
:M-Clarity™ quality level = MQ200
:–24 °C
:GHS02
:GHS07
:Ambient
- Solubility:110 g/l (20 °C)
- Melting Point:-65 °C
- Molar Mass:72.11 g/mol
- Boiling Point:63 - 64.5 °C (1013 mbar)
- Vapor Pressure:189 hPa (20 °C)
- Flash Point:-20 °C
- Explosion Limit:1.6 - 11.0 %(V)
- Density:0.79 g/cm3 (20 °C)
- Ignition Point:165 °C
- Saturation concentration:452 g/m3
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Frequently Bought Together