



n-Pentane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:109-66-0
:
Formula:H₃C(CH₂)₃CH₃
MDL Number:MFCD00009498
:72.15 g/mol
Merck Index:13,07193
Boiling Pt:35…36 °C (1013 hPa)
Melting Pt:–130 °C
Density:0.63 g/cm³ (20 °C)
Flash Pt:–40 °C
EINECS:203-692-4
UN:1265
ADR:3,II
REACH:01-2119459286-30
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Assay:Min. 99 %
- Boiling point:35 - 37 °C
- Density (20/4):0.625 - 0.632
- Evaporation residue:Max. 50 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 30L Metal drum for liquids UN X (ZPACK12124125) 5L Aluminium bottle for liquids DIN 45 UN X (ZPACK8000430)
Packaging Information
spec_VWRC26206.291 spec_VWRC26206.291 spec_VWRC26206.325 spec_VWRC26206.360 spec_VWRC26206.460
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together