



n-Octane 97%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:111-65-9
:
Formula:H₃C(CH₂)₆CH₃
MDL Number:MFCD00009556
:114.23 g/mol
Merck Index:13,06782
Boiling Pt:125…127 °C (760 mmHg)
Melting Pt:–57 °C
Density:0.708 g/cm³
Flash Pt:13 °C
EINECS:203-892-1
UN:1262
ADR:3,II
:GHS07
:GHS08
:GHS09
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3965 to 1.3985 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together