


Sodium dodecyl sulphate (SDS) for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:151-21-3
:
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175
:288.38 g/mol
Merck Index:14,08636
Melting Pt:204…207 °C
Density:1.1 g/cm³ (20 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
UN:1325
ADR:4.1,III
:M-Clarity™ quality level = MQ200
EINECS:205-788-1
:GHS02
:GHS05
:GHS07
Specification Test Results
- Solubility:150 g/l (20 °C)
- Melting Point:204.06 - 207.06 °C
- Molar Mass:288.4 g/mol
- Bulk Density:490 - 560 kg/m3
- Vapor Pressure:(20 °C)
- Flash Point:150 °C
- Density:1.1 g/cm3 (20 °C)
- pH:6 - 9 (10 g/l, H2O, 20 °C)
- Hazard Class:4.1
- Hazard Class Description:Flammable Solid
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC8.22050.9020_TDS_EN ProductPage_822050
Product specification
msds_822050
SDS
Frequently Bought Together