Supelco® Organic Reference Standard, Methanol
82762-10ML-F
: Merck
:
- Pk:10 mL
- Standards:Analytical standard
- CAS No.:67-56-1
- Description:Methanol
- Seq:02
- Storage conditions:2...30 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:STD
- MDL No.:MFCD00004595
- M-Clarity quality level:MQ100
- Synonym(s):Methyl alcohol
- Melting point:–98 °C (lit.)
- Formula:CH₃OH
- Molecular weight:32,04
- Density:0,791 g/ml at 25 °C (lit.)
- Flash point:9,7 °C - Closed cup
- Boiling point:64.7 °C (lit.)
- GHS signal: GHS02, GHS06, GHS08
: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311