Order Entry
Spain
ContactUsLinkComponent
(±)-2-Nonanol for synthesis, Sigma-Aldrich®
(±)-2-Nonanol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
(±)-2-Nonanol for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    -35 °C
  • Molar Mass:
    144.26 g/mol
  • Boiling Point:
    198 °C
  • Flash Point:
    96 °C
  • Refractive Index:
    1.429 (25 °C, 589 nm)
  • Density:
    0.82 g/cm3 (20 °C)
  • Synonyms:
    Heptyl methyl carbinol
  • Hill Formula:
    C9H20O
  • Chemical Formula:
    CH3(CH2)6CH(OH)CH3

Specification Test Results

Related Information

Frequently Bought Together