1-Heptanol ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:111-70-6
:
Formula:H₃C(CH₂)₅CH₂OH
MDL Number:MFCD00002986
:116.2 g/mol
Merck Index:14,4661
Boiling Pt:175…176 °C
Melting Pt:–34 °C
Density:0.820
Flash Pt:73 °C (163 °F)
EINECS:203-897-9
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together