Order Entry
Spain
ContactUsLinkComponent
1-Dodecanol for synthesis, Sigma-Aldrich®
1-Dodecanol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
1-Dodecanol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Melting Point:
    21-24°C
  • Hazard Class:
    9
  • Hazard Class Description:
    Miscellaneous Hazardous Material

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together