

1-Decanol for synthesis, Sigma-Aldrich®
: Merck
:112-30-1
:
:H₃C(CH₂)₈CH₂OH
:MFCD00004747
:158.28 g/mol
:13,02875
:230 °C (1013 hPa)
:7 °C
:0.82634 g/cm³ (20 °C)
:203-956-9
:3082
:9,III
:M-Clarity™ quality level = MQ200
:82 °C
:GHS07
:GHS09
:Ambient
- Solubility:0.037 g/l (25 °C)
- Melting Point:5 - 7 °C
- Molar Mass:158.28 g/mol
- Boiling Point:220 - 235 °C (1013 hPa)
- Vapor Pressure:- 0.1 hPa (20 °C)
- Flash Point:95 °C DIN 51758
- Refractive Index:1.4372 (20 °C, 589 nm)
- Explosion Limit:0.9 - 5.7 %(V)
- Density:0.8 g/cm3 (20 °C) DIN 51757
- Ignition Point:255 °C
- Synonyms:Capric alcohol, Decyl alcohol
- Hill Formula:C10H22O
- Chemical Formula:CH3(CH2)9OH
Frequently Bought Together