

Chloroform ≥99.8% stabilised, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:67-66-3
:
Formula:CHCl₃
MDL Number:MFCD00000826
:119.38 g/mol
Merck Index:13,02160
Boiling Pt:62 °C
Melting Pt:–64 °C
Density:1.49 g/cm³ (20 °C)
Flash Pt: Does not flash
Storage Temperature:Ambient
UN:1888
ADR:6.1,III
:Stabilised with ethanol ~1%
EINECS:200-663-8
REACH:05-2114614906-45
:GHS07
:GHS08
Specification Test Results
- Assay GC (Product + Ethanol):Min. 99.8 %
- Vapor Density:4.1 (vs air)
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.05 %
- Refractive index (n 20/D):1.444 - 1.446
- Suitability for residue analysis:Max. 25 ng/l
- Contains:~1% ethanol as stabilizer
- Vapor Pressure:160 mmHg ( 20 °C)
- Refractive Index:n20/D 1.445 (lit.)
Specification Test Results
Frequently Bought Together