Order Entry
Spain
ContactUsLinkComponent
Chloroform ≥99.5% for HPLC
 
 
 
Chloroform ≥99.5% for HPLC
  22920.K2
 :  
undefined
 
 
 
Chloroform ≥99.5% for HPLC
  22920.K2
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

 :67-66-3
 :
 :CHCl₃
 :MFCD00000826
 :119.38 g/mol
 :14,2141
 :60.5…61.5 °C
 :–63 °C
 :1.492
 :Ambient
 :200-663-8
 :1888
 :6.1,III
 :05-2114614906-45
 :GHS06
 :GHS08
 :GHS07