

Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:75-09-2
:
Formula:CH₂Cl₂
MDL Number:MFCD00000881
:84.93 g/mol
Merck Index:12,06140
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.33 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1593
ADR:6.1,III
:Stabilised with 2-methyl-2-butene (iso-amylene) 20 ppm
EINECS:200-838-9
REACH:01-2119480404-41
:GHS08
:GHS07
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Assay (on anhydrous substance):Min. 99.5 %
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test
- IR Spectrum:Passes test
- Acidity:Max. 0.0003 meq/g
- Boiling point:39 - 40.5 °C
- Colouration:Max. 10 APHA
- Density (20/4):1.322 - 1.327
- Substances coloured by H2SO4:Max. 60 APHA
- Carbon tetrachloride:Max. 100 ppm
- Chloroform:Max. 100 ppm
- Evaporation residue:Max. 10 ppm
- Free halogens:Max. 0.3 ppm
- 2-Methyl-2-butene:10 - 55 ppm
- Water:Max. 0.02 %
- Al (Aluminium):Max. 0.5 ppm
- B (Boron):Max. 0.02 ppm
- Ba (Barium):Max. 0.1 ppm
- Ca (Calcium):Max. 0.5 ppm
- Cd (Cadmium):Max. 0.05 ppm
- Co (Cobalt):Max. 0.02 ppm
- Cr (Chromium):Max. 0.02 ppm
- Cu (Copper):Max. 0.02 ppm
- Fe (Iron):Max. 0.1 ppm
- Mg (Magnesium):Max. 0.1 ppm
- Mn (Manganese):Max. 0.02 ppm
- Ni (Nickel):Max. 0.02 ppm
- Pb (Lead):Max. 0.1 ppm
- Sn (Tin):Max. 0.1 ppm
- Zn (Zinc):Max. 0.1 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249)
Packaging Information
spec_VWRC23366.293 spec_VWRC23366.293 spec_VWRC23366.327 spec_VWRC23366.460 spec_VWRC23366.460
Product specification
More
Frequently Bought Together